Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRP6 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537936
Description
This antibody is predicted to react with mouse, rat, dog and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
The protein encoded by this gene appears to be involved in pre-mRNA splicing, possibly acting as a bridging factor between U5 and U4/U6 snRNPs in formation of the spliceosome. The encoded protein also can bind androgen receptor, providing a link between transcriptional activation and splicing.
Specifications
PRP6 | |
Polyclonal | |
Unconjugated | |
PRPF6 | |
1190003A07Rik; 2610031L17Rik; androgen receptor N-terminal domain transactivating protein-1; androgen receptor N-terminal domain-transactivating protein 1; ANT1; ANT-1; C20orf14; hPrp6; p102 U5 small nuclear ribonucleoprotein particle-binding protein; pre-mRNA processing factor 6; pre-mRNA-processing factor 6; Prp6; PRP6 homolog; PRP6 pre-mRNA processing factor 6 homolog; PRP6 pre-mRNA splicing factor 6 homolog; PRP6 pre-mRNA splicing factor 6 homolog (yeast); Prpf6; putative mitochondrial outer membrane protein import receptor; RGD1307103; RP60; SNRNP102; TOM; U5 snRNP-associated 102 kDa protein; U5-102 kDa protein; U5-102K | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
24148 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O94906 | |
PRPF6 | |
Peptide with sequence C-TGENHTSVDPRQTQ, from the internal region of PRPF6 (aa 193-207). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction