Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRP6 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572186
Description
The protein encoded by this gene appears to be involved in pre-mRNA splicing, possibly acting as a bridging factor between U5 and U4/U6 snRNPs in formation of the spliceosome. The encoded protein also can bind androgen receptor, providing a link between transcriptional activation and splicing.
Specifications
PRP6 | |
Polyclonal | |
Unconjugated | |
PRPF6 | |
ANT-1; ANT1; C20orf14; Prp6; TOM; U5-102K; hPrp6; PRP6 homolog; U5 snRNP-associated 102 kDa protein; U5-102 kDa protein; androgen receptor N-terminal domain transactivating protein-1; androgen receptor N-terminal domain-transactivating protein 1; p102 U5 small nuclear ribonucleoprotein particle-binding protei | |
Rabbit | |
Protein A | |
RUO | |
24148, 68879 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.43 mg/mL | |
PBS with 0.09% sodium azide | |
O94906, Q91YR7 | |
PRPF6 | |
KLH conjugated synthetic peptide between 196-229 amino acids from the N-terminal region of human PRPF6 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction