Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PrPC Rabbit anti-Human, Mouse, Rat, Clone: 3D0J9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PrPC |
---|---|
Clone | 3D0J9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PrPC Monoclonal antibody specifically detects PrPC in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PrPC | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience, Stem Cell Markers | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5621 | |
IgG | |
Affinity purified |
3D0J9 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CD230, CJD, fatal familial insomnia), GSS, prion protein, prion protein (p27-30), prion protein PrP, prion-related protein, PRIPMGC26679 | |
A synthetic peptide corresponding to a sequence within amino acids 154-253 of human PrPC (P04156). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title