Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPF18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537022
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537022 Supplier Invitrogen™ Supplier No. PA537022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 40 kDa. Purity is >95% by SDS-PAGE.

PRPF18 (pre-mRNA-splicing factor 18), also known as HPRP18, is a 342 amino acid protein that localizes to nuclear speckles and plays a role in the second step of pre-mRNA splicing. A member of the PRP18 family, PRPF18 contains seven WD repeats and exists as two alternatively spliced isoforms which are encoded by a gene located on human chromosome 10p13. Chromosome 10 contains over 800 genes and 135 million nucleotides. PTEN is an important tumor suppressor gene located on chromosome 10 and, when defective, causes a genetic predisposition to cancer development known as Cowden syndrome. Other chromosome 10 associated disorders include Cockayne syndrome, tetrahydrobiopterin deficiency and trisomy 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF18
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene PRPF18
Gene Accession No. Q8BM39, Q99633, Q9JKB8
Gene Alias 2810441A10Rik; HPRP18; HPRP18 homolog; Kcrf; Potassium channel regulatory factor; Pprf18; pre-mRNA processing factor 18; pre-mRNA-splicing factor 18; Prp18; PRP18 homolog; PRP18 pre-mRNA processing factor 18 homolog; PRP18 pre-mRNA processing factor 18 homolog (yeast); PRP18 pre-mRNA processing factor 18 homolog(PRPF18); Prpf18
Gene Symbols PRPF18
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 223-268 of human PRPF18.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171552, 67229, 8559
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.