Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRPF3 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119114V
Description
PRPF3 Polyclonal antibody specifically detects PRPF3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PRPF3 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
hPrp3, HPRP3P, HPRP3retinitis pigmentosa 18 (autosomal dominant), Pre-mRNA-splicing factor 3, PRP3, PRP3 pre-mRNA processing factor 3 homolog (S. cerevisiae), PRP3 pre-mRNA processing factor 3 homolog (yeast), Prp3p, RP18, U4/U6 small nuclear ribonucleoprotein Prp3, U4/U6 snRNP 90 kDa protein, U4/U6-associated RNA splicing factor | |
The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human U4/U6 small nuclear ribonucleoprotein Prp3 using the numbering given in entry NP_004689.1 (GeneID 9129). | |
0.1 mL | |
Vision | |
9129 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction