Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102783 Supplier Invitrogen™ Supplier No. PA5102783
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PRPF3.

The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF3 is 1 of several proteins that associate with U4 and U6 snRNPs. The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF3 is 1 of several proteins that associate with U4 and U6 snRNPs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRPF3
Gene Accession No. O43395, Q922U1
Gene Alias 3632413F13Rik; HPRP3; HPRP3P; MGC137653 protein; pre-mRNA processing factor 3; pre-mRNA-splicing factor 3; PRP3; PRP3 pre-mRNA processing factor 3 homolog; PRP3 pre-mRNA processing factor 3 homolog (yeast); Prp3p; PRPF3; RP18; SNRNP90; U4/U6 small nuclear ribonucleoprotein Prp3; U4/U6 snRNP 90 kDa protein; U4/U6-associated RNA splicing factor
Gene Symbols PRPF3
Host Species Rabbit
Immunogen A synthesized peptide derived from human PRPF3(Accession O43395), corresponding to amino acid residues P53-K103.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361995, 70767, 9129
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.