Learn More
PRPF3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Shop All Proteintech Group Inc Products

Description
This antibody is a rabbit polyclonal antibody raised against an internal region of human PRPF3. A predicted molecular mass of 77 kDa and the gene is named PRPF3 because it is homologous to the yeast PRP3 gene. We noticed that although the predicted molecu
Specifications
Specifications
| Antigen | PRPF3 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | PRPF3 |
| Gene Accession No. | O43395, Q922U1 |
| Gene Alias | HPRP3, HPRP3P, Pre mRNA splicing factor 3, PRP3, Prp3p, PRPF3, RP18, U4/U6 snRNP 90 kDa protein |
| Gene Symbols | Prpf3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.