Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRPF31 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | PRPF31 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PRPF31 Polyclonal specifically detects PRPF31 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
PRPF31 | |
Polyclonal | |
Rabbit | |
Human | |
Q8WWY3 | |
26121 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TNATIMVVSVTASTTQGQQLSEEELERLEEACDMALELNASKHRIYEYVESRMSFIAPNLSIIIGASTAAKIMGVAGGLTNLSKMPACNIMLLGAQRKTLSGF | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
55 kDa |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DKFZp566J153, hPrp31, NY-BR-99, Pre-mRNA-processing factor 31, Protein 61K, PRP31 pre-mRNA processing factor 31 homolog (S. cerevisiae), PRP31 pre-mRNA processing factor 31 homolog (yeast), PRP31RP11, Serologically defined breast cancer antigen NY-BR-99, U4/U6 small nuclear ribonucleoprotein Prp31, U4/U6 snRNP 61 kDa protein | |
PRPF31 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title