Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPF31 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110222
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110222 Supplier Invitrogen™ Supplier No. PA5110222
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRPF31 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a component of the spliceosome complex and is one of several retinitis pigmentosa-causing genes. When the gene product is added to the spliceosome complex, activation occurs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF31
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene PRPF31
Gene Accession No. Q8CCF0, Q8WWY3
Gene Alias 1500019O16Rik; 2810404O06Rik; AW554706; DKFZp566J153; hPrp 31; hPrp31; NY BR 99; NY-BR-99; pre-mRNA processing factor 31; pre-mRNA-processing factor 31; Protein 61K; PRP 31; Prp31; PRP31 pre-mRNA processing factor 31 homolog; PRP31 pre-mRNA processing factor 31 homolog (yeast); PRPF 31; Prpf31; RP 11; RP11; serologically defined breast cancer antigen NY-BR-99; SNRNP61; U4/U6 small nuclear ribonucleoprotein Prp31; U4/U6 snRNP 61 kDa protein
Gene Symbols PRPF31
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26121, 292536, 68988
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.