Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRPF31 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576808
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a component of the spliceosome complex and is one of several retinitis pigmentosa-causing genes. When the gene product is added to the spliceosome complex, activation occurs.
Specifications
PRPF31 | |
Polyclonal | |
Unconjugated | |
PRPF31 | |
1500019O16Rik; 2810404O06Rik; AW554706; DKFZp566J153; hPrp 31; hPrp31; NY BR 99; NY-BR-99; pre-mRNA processing factor 31; pre-mRNA-processing factor 31; Protein 61K; PRP 31; Prp31; PRP31 pre-mRNA processing factor 31 homolog; PRP31 pre-mRNA processing factor 31 homolog (yeast); PRPF 31; Prpf31; RP 11; RP11; serologically defined breast cancer antigen NY-BR-99; SNRNP61; U4/U6 small nuclear ribonucleoprotein Prp31; U4/U6 snRNP 61 kDa protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26121, 292536, 68988 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8CCF0, Q8WWY3 | |
PRPF31 | |
Recombinant full length Human PRPF31. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction