Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRPF31 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583581
Description
Immunogen sequence: TNATIMVVSV TASTTQGQQL SEEELERLEE ACDMALELNA SKHRIYEYVE SRMSFIAPNL SIIIGASTAA KIMGVAGGLT NLSKMPACNI MLLGAQRKTL SGF.
This gene encodes a component of the spliceosome complex and is one of several retinitis pigmentosa-causing genes. When the gene product is added to the spliceosome complex, activation occurs.
Specifications
| PRPF31 | |
| Polyclonal | |
| Unconjugated | |
| PRPF31 | |
| 1500019O16Rik; 2810404O06Rik; AW554706; DKFZp566J153; hPrp 31; hPrp31; NY BR 99; NY-BR-99; pre-mRNA processing factor 31; pre-mRNA-processing factor 31; Protein 61K; PRP 31; Prp31; PRP31 pre-mRNA processing factor 31 homolog; PRP31 pre-mRNA processing factor 31 homolog (yeast); PRPF 31; Prpf31; RP 11; RP11; serologically defined breast cancer antigen NY-BR-99; SNRNP61; U4/U6 small nuclear ribonucleoprotein Prp31; U4/U6 snRNP 61 kDa protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26121 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.08 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WWY3 | |
| PRPF31 | |
| Recombinant protein corresponding to Human PRPF31. Recombinant protein control fragment (Product #RP-98980). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction