Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPF31 Recombinant Rabbit Monoclonal Antibody (JE64-82)
GREENER_CHOICE

Catalog No. PIMA544719
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544719 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544719 Supplier Invitrogen™ Supplier No. MA544719
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

PRPF31 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

This gene encodes a component of the spliceosome complex and is one of several retinitis pigmentosa-causing genes. When the gene product is added to the spliceosome complex, activation occurs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF31
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE64-82
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PRPF31
Gene Accession No. Q8WWY3
Gene Alias 1500019O16Rik; 2810404O06Rik; AW554706; DKFZp566J153; hPrp 31; hPrp31; NY BR 99; NY-BR-99; pre-mRNA processing factor 31; pre-mRNA-processing factor 31; Protein 61K; PRP 31; Prp31; PRP31 pre-mRNA processing factor 31 homolog; PRP31 pre-mRNA processing factor 31 homolog (yeast); PRPF 31; Prpf31; RP 11; RP11; serologically defined breast cancer antigen NY-BR-99; SNRNP61; U4/U6 small nuclear ribonucleoprotein Prp31; U4/U6 snRNP 61 kDa protein
Gene Symbols PRPF31
Host Species Rabbit
Immunogen Synthetic peptide within human PRPF31 aa 350-399/499.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26121
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.