Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRPF38A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP237697
Description
PRPF38A Polyclonal specifically detects PRPF38A in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
PRPF38A | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
FLJ14936, MGC3320, pre-mRNA-splicing factor 38A, Prp38, PRP38 pre-mRNA processing factor 38 (yeast) domain containing A, RP5-965L7.1 | |
Rabbit | |
Affinity Purified | |
RUO | |
84950 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate | |
NP_116253.2 | |
PRPF38A | |
This antibody maps to a region between residue 262 to 312 of human Pre-mRNA Processing Factor 38A using the numbering given in entry NP_116253.2 (GeneID 84950). | |
100 μL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction