Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPF38A Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501568255
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8255 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8255 Supplier Bethyl Laboratories, Inc Supplier No. A304635AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with X. laevis, X. tropicalis, Bovine and Orangutan

PRPF38A is a protein coding gene involved in pre-mRNA splicing as a component of the spliceosome.

Specifications

Antigen PRPF38A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PRPF38A
Gene Accession No. Q4FK66, Q8NAV1
Gene Alias Prp38, RP5-965L7.1, pre-mRNA-splicing factor 38A
Gene Symbols PRPF38A
Host Species Rabbit
Immunogen Between 262 and 312
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230596, 84950
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.