Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPF39 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551528
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551528 Supplier Invitrogen™ Supplier No. PA551528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DINWENEQGN LREVTAIYDR ILGIPTQLYS HHFQRFKEHV QNNLPRDLLT GEQFIQLRRE LASVNGHSGD DGPPGDDLPS GIEDITDPAK LITEIENMRH RIIEIHQEMF NYNEHEVSKR WTFEEGIKRP YFHVKPLEKA QL Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.

The PRPF39 gene plays a critical role as a splicing factor involved in alternative splicing regulation. In humans, it is homologous to the yeast Prp39 and Prp42 paralogs, forming a homodimer that interacts with the carboxy-terminal domain (CTD) of U1C, a parallel mechanism observed in the yeast Prp39/Prp42 heterodimer. Recent studies have highlighted its impact on splicing efficiency through knockdown experiments in HEK293 cells, which revealed significant changes in the expression of many genes, underscoring its importance in splicing adaptability according to cell type-specific needs. Additionally, PRPF39 is found to be highly expressed in Acute Lymphoblastic Leukemia (ALL), with research suggesting that targeting PRPF39 for degradation could contribute to the treatment of ALL. Phylogenetic analyses indicate a coevolution of PRPF39 expression with molecular components like shortened U1 snRNA, reflecting the splicing complexity across different species. The gene is also involved in the regulation of distant genes which are relevant to cellular responses, such as those involved in cisplatin response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF39
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRPF39
Gene Accession No. Q86UA1, Q8K2Z2
Gene Alias flj11128; flj20666; flj45460; mgc149842; mgc149843; pre-mRNA processing factor 39; pre-mRNA-processing factor 39; PRP39 homolog; PRP39 pre-mRNA processing factor 39 homolog; PRP39 pre-mRNA processing factor 39 homolog (yeast); PRPF39; Srcs1; v-src suppressed transcript 1
Gene Symbols PRPF39
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRPF39. Recombinant protein control fragment (Product #RP-89011).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314171, 328110, 55015
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.