Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541794
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541794 Supplier Invitrogen™ Supplier No. PA541794
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EVFEIEEHIS ERQAEVLAEF ERRKRARQIN VSTDDSEVKA CLRALGEPIT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF4 is 1 of several proteins that associate with U4 and U6 snRNPs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRPF4
Gene Accession No. O43172
Gene Alias 1600015H11Rik; AI874830; AW047464; bN189G18.1; HPRP4; HPRP4P; hypothetical protein LOC508735; mg:ab03a02; pre-mRNA processing factor 4; PRP4; PRP4 homolog; PRP4 pre-mRNA processing factor 4 homolog; PRP4 pre-mRNA processing factor 4 homolog (yeast); PRP4/STK/WD splicing factor; Prp4p; PRPF4; RP70; SNRNP60; U4/U6 small nuclear ribonucleoprotein Prp4; U4/U6 snRNP 60 kDa protein; WD splicing factor Prp4; zgc:65943
Gene Symbols PRPF4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PRPF4 (aa 71-120).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9128
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.