Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPF4 Recombinant Rabbit Monoclonal Antibody (JE65-37)
GREENER_CHOICE

Catalog No. PIMA544745
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544745 Supplier Invitrogen™ Supplier No. MA544745
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

PRPF4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF4 is 1 of several proteins that associate with U4 and U6 snRNPs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE65-37
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PRPF4
Gene Accession No. O43172
Gene Alias 1600015H11Rik; AI874830; AW047464; bN189G18.1; HPRP4; HPRP4P; hypothetical protein LOC508735; mg:ab03a02; pre-mRNA processing factor 4; PRP4; PRP4 homolog; PRP4 pre-mRNA processing factor 4 homolog; PRP4 pre-mRNA processing factor 4 homolog (yeast); PRP4/STK/WD splicing factor; Prp4p; PRPF4; RP70; SNRNP60; U4/U6 small nuclear ribonucleoprotein Prp4; U4/U6 snRNP 60 kDa protein; WD splicing factor Prp4; zgc:65943
Gene Symbols PRPF4
Host Species Rabbit
Immunogen Synthetic peptide within human PRPF4 aa 1-50/522.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9128
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.