Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRPF40A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558320
Description
Immunogen sequence: NASTSASNTV SGTVPVVPEP EVTSIVATVV DNENTVTIST EEQAQLTSTP AIQDQSVEVS SNTGEETSKQ ETVADFTP Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 73%.
FNBP3, Formin-binding protein 3, is a 957 amino acid protein encoded by the human gene PRPF40A. FNBP3 belongs to the PRPF40 family and contains five FF domains and two WW domains. Nuclear proteins harboring both WW and FF protein interaction modules bind to splicing factors as well as RNA polymerase II and may serve to link transcription with splicing. Through the WW domains, FNBP3 will bind to the Formin proline-rich regions. Also known as Pre-mRNA-processing factor 40 homolog A, FNBP3 binds to WASL/N-WASP (Neuronal Wiskott-Aldrich syndrome protein) complex and suppresses its translocation from the nucleus to the cytoplasm, thereby inhibiting its cytoplasmic function. FNBP3 is widely expressed in most tissues and is localized to the nuclear speckles.
Specifications
PRPF40A | |
Polyclonal | |
Unconjugated | |
PRPF40A | |
2810012K09Rik; Fas ligand-associated factor 1; Fas-ligand associated factor 1; FBP11; FBP-11; FLAF1; FLJ20585; Fnbp11; Fnbp3; formin binding protein 11; formin binding protein 3; formin-binding protein 11; Formin-binding protein 3; HIP10; HIP-10; HSPC225; Huntingtin yeast partner A; huntingtin-interacting protein 10; Huntingtin-interacting protein A; HYPA; NY-REN-6; NY-REN-6 antigen; pre-mRNA processing factor 40 homolog A; pre-mRNA-processing factor 40 homolog A; Prp40; PRP40 pre-mRNA processing factor 40 homolog A; PRP40 pre-mRNA processing factor 40 homolog A (S. cerevisiae); PRP40 pre-mRNA processing factor 40 homolog A (yeast); PRPF40A; Renal carcinoma antigen NY-REN-6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55660 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75400 | |
PRPF40A | |
Recombinant protein corresponding to Human PRPF40A. Recombinant protein control fragment (Product #RP-96227). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction