Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRPF40A Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PRPF40A |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1547
![]() ![]() |
Proteintech Group Inc
173921AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
FNBP3, Formin-binding protein 3, is a 957 amino acid protein encoded by the human gene PRPF40A. FNBP3 belongs to the PRPF40 family and contains five FF domains and two WW domains. Nuclear proteins harboring both WW and FF protein interaction modules bind to splicing factors as well as RNA polymerase II and may serve to link transcription with splicing. Through the WW domains, FNBP3 will bind to the Formin proline-rich regions. Also known as Pre-mRNA-processing factor 40 homolog A, FNBP3 binds to WASL/N-WASP (Neuronal Wiskott-Aldrich syndrome protein) complex and suppresses its translocation from the nucleus to the cytoplasm, thereby inhibiting its cytoplasmic function. FNBP3 is widely expressed in most tissues and is localized to the nuclear speckles.Specifications
PRPF40A | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O75400, Q9R1C7 | |
55660, 56194 | |
PRPF40A Fusion Protein Ag11290 | |
Primary | |
-20°C | |
PRPF40A |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Fas ligand associated factor 1, FBP 11, FBP11, FLAF1, FNBP3, Formin binding protein 11, Formin binding protein 3, HIP 10, HIP10, Huntingtin yeast partner A, HYPA, NY REN 6, PRPF40A | |
PRPF40A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title