Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPF40B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558461 Supplier Invitrogen™ Supplier No. PA558461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PQPQPDPPPV PPGPTPVPTG LLEPEPGGSE DCDVLEATQP LEQGFLQQLE EGPSSSGQHQ PQQEEEESKP EPERSGLSWS NREKAKQAF Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 83%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF40B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRPF40B
Gene Accession No. Q6NWY9
Gene Alias 2610317D23Rik; Huntingtin interacting protein C; huntingtin yeast partner C; Huntingtin-interacting protein C; huntington yeast partner C; Hypc; pre-mRNA processing factor 40 homolog B; pre-mRNA-processing factor 40 homolog B; PRP40 pre-mRNA processing factor 40 homolog A; PRP40 pre-mRNA processing factor 40 homolog B; PRP40 pre-mRNA processing factor 40 homolog B (S. cerevisiae); PRP40 pre-mRNA processing factor 40 homolog B (yeast); Prpf40b; RGD1565411
Gene Symbols PRPF40B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRPF40B. Recombinant protein control fragment (Product #RP-97672).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25766
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.