Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPF4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571896
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571896 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571896 Supplier Invitrogen™ Supplier No. PA571896
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRPF4B Polyclonal Antibody for Western Blot

Pre-mRNA splicing occurs in two sequential transesterification steps, and the protein encoded by this gene is thought to be involved in pre-mRNA splicing and in signal transduction. This protein belongs to a kinase family that includes serine/arginine-rich protein-specific kinases and cyclin-dependent kinases. This protein is regarded as a CDK-like kinase with homology to mitogen-activated protein kinases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF4B
Applications Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PRPF4B
Gene Accession No. Q61136
Gene Alias 2610037H07; cbp143; dJ1013A10.1; dJ1013A10.1 (PRP4 protein kinase homolog); KIAA0536; PR4H; pre-mRNA processing factor 4B; pre-mRNA protein kinase; protein-serine/threonine kinase; Prp4; PRP4 kinase; PRP4 pre-mRNA processing factor 4 homolog B; PRP4 pre-mRNA processing factor 4 homolog B (yeast); PRP4 pre-mRNA-processing factor 4 homolog; PRP4H; PRP4K; Prp4m; Prpf4b; Prpk; RP5-1013A10.2; Serine/threonine-protein kinase PRP4 homolog
Gene Symbols PRPF4B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 180-207 amino acids from the N-terminal region of mouse Prpf4b.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19134
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.