Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPF4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110223
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110223 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110223 Supplier Invitrogen™ Supplier No. PA5110223
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QKYKYLAEDS NMSVPSEPSS PQSSTRTRSP SPDDILERVA ADVKEYEREN VDTFEASVKA KHNLMTVEQN NGSSQKKLLA PDMFTESDDM FAAYFDSARL RAAGIGKDFK ENPNLRDNWT DAEGYYRVNI GEVLDKRYNV YGYTGQGVFS NVVRARDNAR ANQEVAVKII RNNELMQKTG LKELEFLKKL NDADPDDKFH CLRLFRHFYH K.

Pre-mRNA splicing occurs in two sequential transesterification steps, and the protein encoded by this gene is thought to be involved in pre-mRNA splicing and in signal transduction. This protein belongs to a kinase family that includes serine/arginine-rich protein-specific kinases and cyclin-dependent kinases. This protein is regarded as a CDK-like kinase with homology to mitogen-activated protein kinases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPF4B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PRPF4B
Gene Accession No. Q13523, Q5RKH1, Q61136
Gene Alias 2610037H07; cbp143; dJ1013A10.1; dJ1013A10.1 (PRP4 protein kinase homolog); KIAA0536; PR4H; pre-mRNA processing factor 4B; pre-mRNA protein kinase; protein-serine/threonine kinase; Prp4; PRP4 kinase; PRP4 pre-mRNA processing factor 4 homolog B; PRP4 pre-mRNA processing factor 4 homolog B (yeast); PRP4 pre-mRNA-processing factor 4 homolog; PRP4H; PRP4K; Prp4m; Prpf4b; Prpk; RP5-1013A10.2; Serine/threonine-protein kinase PRP4 homolog
Gene Symbols PRPF4B
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19134, 291078, 8899
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.