Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPF8 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501566413
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6413 Supplier Bethyl Laboratories, Inc Supplier No. A303920A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Pre-mRNA splicing occurs in 2 sequential transesterification steps. The protein encoded by this gene is a component of both U2- and U12-dependent spliceosomes, and found to be essential for the catalytic step II in pre-mRNA splicing process. It contains several WD repeats, which function in protein-protein interactions. This protein has a sequence similarity to yeast Prp8 protein. This gene is a candidate gene for autosomal domit retinitis pigmentosa.

Specifications

Antigen PRPF8
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PRPF8
Gene Accession No. Q6P2Q9, Q99PV0
Gene Alias HPRP8, PRP8, PRPC8, RP13, 220 kDa U5 snRNP-specific protein, PRP8 homolog, U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p, apoptosis-regulated protein 1, apoptosis-regulated protein 2, p220, pre-mRNA-processing-splicing factor 8, precursor mRNA processing protein, spl
Gene Symbols Prpf8
Host Species Rabbit
Immunogen Between 850 and 900
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10594, 192159
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.