Learn More
PRPF8 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303921A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Pre-mRNA splicing occurs in 2 sequential transesterification steps. The protein encoded by this gene is a component of both U2- and U12-dependent spliceosomes, and found to be essential for the catalytic step II in pre-mRNA splicing process. It contains several WD repeats, which function in protein-protein interactions. This protein has a sequence similarity to yeast Prp8 protein. This gene is a candidate gene for autosomal domit retinitis pigmentosa.Specifications
PRPF8 | |
Polyclonal | |
Unconjugated | |
PRPF8 | |
HPRP8, PRP8, PRPC8, RP13, 220 kDa U5 snRNP-specific protein, PRP8 homolog, U5 snRNP-specific protein (220 kD), ortholog of S. cerevisiae Prp8p, apoptosis-regulated protein 1, apoptosis-regulated protein 2, p220, pre-mRNA-processing-splicing factor 8, precursor mRNA processing protein, spl | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10594, 192159 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q6P2Q9, Q99PV0 | |
Prpf8 | |
Between 1825 and 1875 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.