Learn More
PRPH Mouse anti-Human, Clone: OTI8G3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806240

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Peripherin is a 57kD type III intermediate filament that is a specific marker for peripheral neurons, including enteric ganglion cells. Peripherin is expressed in the developing peripheral nervous system and is highly enriched in neuronal derivatives of the neural crest. Peripherin offers an advantage over other neural markers, such as S100 and NSE in that it does not stain chrmaffin cell types.Specifications
PRPH | |
Monoclonal | |
Unconjugated | |
PRPH | |
NEF4, PRPH1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5630 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8G3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P41219 | |
PRPH | |
Full length human recombit protein of human PRPH produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.