Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPH2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981160
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1160 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1160 Supplier Bioss Supplier No. BS11197R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peripherin-2 (PRPH2), also known as retinal degeneration slow protein (RDS), is a photoreceptor-specific tetraspanin protein implicated in outer segment disk morphogenesis. It may function as an adhesion molecule involved in stabilization and compaction of outer segment disks or in the maintenance of the curvature of the rim. Mutations in peripherin-2 are responsible for various retinal degenerative diseases including autosomal dominant retinitis pigmentosa (ADRP).

Specifications

Antigen PRPH2
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PRPH2
Gene Accession No. P23942
Gene Alias AOFMD; AVMD; CACD2; DS; MDBS1; Nmf193; peripherin 2; peripherin 2 (retinal degeneration, slow); peripherin 2, homolog of mouse; peripherin, photoreceptor type; Peripherin-2; PRPH; PRPH2; Rd2; Rd-2; Rds; retinal degeneration 2; Retinal degeneration slow protein; retinal degeneration, slow; retinal degeneration, slow (retinitis pigmentosa 7); retinal peripherin; RP7; RSRDS; Tetraspanin-22; TSPAN22; Tspan-22
Gene Symbols PRPH2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PRPH2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5961
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.