Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPK Monoclonal Antibody (OTI5A5)
GREENER_CHOICE

Catalog No. PIMA526956
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526956 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526956 Supplier Invitrogen™ Supplier No. MA526956
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRPK Monoclonal Antibody for Western Blot, IHC (P)

Component of the EKC/KEOPS complex that is required for the formation of a threonylcarbamoyl group on adenosine at position 37 (t(6)A37) in tRNAs that read codons beginning with adenine (PubMed:22912744, PubMed:27903914). The complex is probably involved in the transfer of the threonylcarbamoyl moiety of threonylcarbamoyl-AMP (TC-AMP) to the N6 group of A37 (PubMed:22912744, PubMed:27903914). TP53RK has ATPase activity in the context of the EKC/KEOPS complex and likely plays a supporting role to the catalytic subunit OSGEP. Atypical protein kinase that phosphorylates 'ser-15' of p53/TP53 protein and may therefore participate in its activation (PubMed:11546806). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5A5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Tp53rk
Gene Accession No. Q96S44
Gene Alias 2810408M09Rik; 4933401B08Rik; 5630401H01Rik; Atypical serine/threonine protein kinase TP53RK; BUD32; C20orf64; dJ101A2; EKC/KEOPS complex subunit TP53RK; mNori-2p; Nori-2; Nori-2p; p53-related protein kinase; PRPK; TP53 regulating kinase; TP53-regulating kinase; TP53RK; transformation related protein 53 regulating kinase; Trp53 regulating kinase; Trp53rk; Trp53tk
Gene Symbols Tp53rk
Host Species Mouse
Immunogen Full length human recombinant protein of TP53RK produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112858
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.