Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPK Monoclonal Antibody (OTI5B2)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526957

Catalog No. PIMA526957


Only null left
Add to Cart

Description

Description

PRPK Monoclonal Antibody for Western Blot, IHC (P)

Component of the EKC/KEOPS complex that is required for the formation of a threonylcarbamoyl group on adenosine at position 37 (t(6)A37) in tRNAs that read codons beginning with adenine (PubMed:22912744, PubMed:27903914). The complex is probably involved in the transfer of the threonylcarbamoyl moiety of threonylcarbamoyl-AMP (TC-AMP) to the N6 group of A37 (PubMed:22912744, PubMed:27903914). TP53RK has ATPase activity in the context of the EKC/KEOPS complex and likely plays a supporting role to the catalytic subunit OSGEP. Atypical protein kinase that phosphorylates 'Ser-15' of p53/TP53 protein and may therefore participate in its activation (PubMed:11546806). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PRPK
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q96S44
Tp53rk
Full length human recombinant protein of TP53RK produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Immunohistochemistry (Paraffin), Western Blot
OTI5B2
Unconjugated
Tp53rk
2810408M09Rik; 4933401B08Rik; 5630401H01Rik; Atypical serine/threonine protein kinase TP53RK; BUD32; C20orf64; dJ101A2; EKC/KEOPS complex subunit TP53RK; mNori-2p; Nori-2; Nori-2p; p53-related protein kinase; PRPK; TP53 regulating kinase; TP53-regulating kinase; TP53RK; transformation related protein 53 regulating kinase; Trp53 regulating kinase; Trp53rk; Trp53tk
Mouse
Affinity Chromatography
RUO
112858
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.