Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPS1 Antibody (1E11) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242649

Catalog No. NBP242649


Only null left
Add to Cart

Description

Description

Ensure accurate, reproducible results in Western Blot, ELISA

PRPS1 Monoclonal specifically detects PRPS1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PRPS1
Monoclonal
1 mg/mL
Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100
NP_002755
PRPS1
Recombinant human PRPS1 (1-318aa) purified from E. coli
100 μL
Apoptosis, Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers
5631
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG2a κ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
100000000000
Unconjugated
PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide
CMTX5ARTS, deafness, X-linked 2, perceptive, congenital, DFN2, DFNX1KIAA0967, dJ1070B1.2 (phosphoribosyl pyrophosphate synthetase 1), EC 2.7.6.1, Phosphoribosyl pyrophosphate synthase I, phosphoribosyl pyrophosphate synthetase 1, PPRibP, PRSI, PRS-I, ribose-phosphate diphosphokinase 1, ribose-phosphate pyrophosphokinase 1
Mouse
Protein A purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only