Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560499
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560499 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560499 Supplier Invitrogen™ Supplier No. PA560499
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LHASQIQVSV EAKYWCLKGG RKGLMMLKTA EKP Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 39%.

This gene encodes an enzyme that catalyzes the phosphoribosylation of ribose 5-phosphate to 5-phosphoribosyl-1-pyrophosphate, which is necessary for purine metabolism and nucleotide biosynthesis. Defects in this gene are a cause of phosphoribosylpyrophosphate synthetase superactivity, Charcot-Marie-Tooth disease X-linked recessive type 5 and Arts Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPS1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Prps1
Gene Accession No. P60891
Gene Alias 2310010D17Rik; ARTS; C76571; C76678; CMTX5; deafness 2, perceptive, congenital; deafness, X-linked 2, perceptive, congenital; DFN2; DFNX1; dJ1070B1.2 (phosphoribosyl pyrophosphate synthetase 1); phosphoribosyl pyrophosphate synthase I; phosphoribosyl pyrophosphate synthetase 1; phosphoribosyl pyrophosphate synthetase I; PPRibP; PRPS1; Prps-1; PRSI; PRS-I; ribose-phosphate diphosphokinase 1; ribose-phosphate pyrophosphokinase 1
Gene Symbols Prps1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRPS1. Recombinant protein control fragment (Product #RP-100426).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5631
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.