Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117121
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117121 Supplier Thermo Scientific Supplier No. PA5117121
Add to Cart
Add to Cart

PRPS1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPS1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Prps1
Gene Accession No. P60891, P60892, Q9D7G0
Gene Alias 2310010D17Rik; ARTS; C76571; C76678; CMTX5; deafness 2, perceptive, congenital; deafness, X-linked 2, perceptive, congenital; DFN2; DFNX1; dJ1070B1.2 (phosphoribosyl pyrophosphate synthetase 1); phosphoribosyl pyrophosphate synthase I; phosphoribosyl pyrophosphate synthetase 1; phosphoribosyl pyrophosphate synthetase I; PPRibP; PRPS1; Prps-1; PRSI; PRS-I; ribose-phosphate diphosphokinase 1; ribose-phosphate pyrophosphokinase 1
Gene Symbols Prps1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-318 of human PRPS1 (NP_002755.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19139, 29562, 5631
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.