Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRPS1/PRPS2/PRPS3 Rabbit anti-Human, Mouse, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB30838
Description
Rabbit polyclonal antibody raised against synthetic peptide of PRPS1/2/3.
- High specificity
- High reactivity
Specifications
PRPS1/PRPS2/PRPS3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of PRPS1/2/3. | |
In PBS (0.09% sodium azide) | |
ARTS/CMTX5/KIAA0967/PPRibP/PRSI | |
Rabbit | |
Saturated Ammonium Sulfate precipitation | |
RUO | |
5631/5634/221823 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
P60891 | |
PRPS1 | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human PRPS1/2/3. | |
400 μL | |
Primary | |
Human, Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction