Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPS1L1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP133410

Catalog No. NBP133410


Only null left
Add to Cart

Description

Description

PRPS1L1 Polyclonal specifically detects PRPS1L1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PRPS1L1
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
PRPS1L1
Full length human PRPS1L1 Recombinant protein.
Affinity Purified
RUO
Primary
Tested in human. Not yet tested in any other species.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot, Immunocytochemistry, Immunofluorescence
1.0mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
EC 2.7.6.1, Phosphoribosyl pyrophosphate synthase III, phosphoribosyl pyrophosphate synthetase 1-like 1, phosphoribosylpyrophosphate synthetase subunit III, PRPS1, PRPS1-like 1, PRPS3Phosphoribosyl pyrophosphate synthase 1-like 1, PRPSL, PRS-III, ribose-phosphate diphosphokinase catalytic chain III, ribose-phosphate pyrophosphokinase 3, ribose-phosphate pyrophosphokinase III
Rabbit
35 kDa
100 μL
Lipid and Metabolism
221823
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only