Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPS1L1 Monoclonal Antibody (OTI2C6)
GREENER_CHOICE

Catalog No. PIMA527509
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527509 Supplier Invitrogen™ Supplier No. MA527509
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRPS1L1 Monoclonal Antibody for Western Blot

Catalyzes the synthesis of phosphoribosylpyrophosphate (PRPP) that is essential for nucleotide synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPS1L1
Applications Western Blot
Classification Monoclonal
Clone OTI2C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRPS1L1
Gene Accession No. P21108
Gene Alias 1700011K15Rik; hCG_1643027; phosphoribosyl pyrophosphate synthase 1-like 1; Phosphoribosyl pyrophosphate synthase III; phosphoribosyl pyrophosphate synthetase 1-like 1; phosphoribosylpyrophosphate synthetase subunit III; PRPS1; Prps1l1; PRPS1-like 1; PRPS3; PRPSL; PRS-III; ribose-phosphate diphosphokinase catalytic chain III; ribose-phosphate pyrophosphokinase 3; ribose-phosphate pyrophosphokinase III
Gene Symbols PRPS1L1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2-318 of PRPS1L1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221823
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.