Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPS1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116324
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116324 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116324 Supplier Invitrogen™ Supplier No. PA5116324
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRPS1L1 Polyclonal Antibody for Western Blot

Catalyzes the synthesis of phosphoribosylpyrophosphate (PRPP) that is essential for nucleotide synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPS1L1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PRPS1L1
Gene Accession No. P21108
Gene Alias 1700011K15Rik; hCG_1643027; phosphoribosyl pyrophosphate synthase 1-like 1; Phosphoribosyl pyrophosphate synthase III; phosphoribosyl pyrophosphate synthetase 1-like 1; phosphoribosylpyrophosphate synthetase subunit III; PRPS1; Prps1l1; PRPS1-like 1; PRPS3; PRPSL; PRS-III; ribose-phosphate diphosphokinase catalytic chain III; ribose-phosphate pyrophosphokinase 3; ribose-phosphate pyrophosphokinase III
Gene Symbols PRPS1L1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 77-106 amino acids from the N-terminal region of human PRPS1L1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221823
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.