Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRPSAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590638
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590638 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590638 Supplier Invitrogen™ Supplier No. PA590638
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNAARTGYRV FSANSTAACT ELAKRITERL GAELGKSVVY QETNGETRVE IKESVRGQDI FIIQTIPRDV NTAVMELLIM A; Positive Samples: HeLa, Mouse heart.

Seems to play a negative regulatory role in 5-phosphoribose 1-diphosphate synthesis. Target protein binds catalytic subunits of the ribose-phosphate diphosphokinase enzyme complex; may act as a negative regulator of enzyme activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRPSAP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PRPSAP1
Gene Accession No. Q14558, Q9D0M1
Gene Alias 39 kDa phosphoribosypyrophosphate synthase-associated protein; 39 kDa phosphoribosypyrophosphate synthetase-associated protein; 5730409F23Rik; hypothetical protein LOC431746; kpra; PAP39; Phosphoribosyl pyrophosphate synthase-associated protein 1; phosphoribosyl pyrophosphate synthetase associated protein 1; phosphoribosyl pyrophosphate synthetase-associated protein 1; phosphoribosylpyrophosphate synthetase-associated protein (39 kDa); PRPP synthase-associated protein 1; PRPP synthetase-associated protein 1; Prpsap1; zgc:91845
Gene Symbols PRPSAP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-110 of human PRPSAP1 (NP_0027572).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5635, 67763
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.