Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRPSAP2 Antibody (OTI1E3), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273676AF532
Description
PRPSAP2 Monoclonal specifically detects PRPSAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PRPSAP2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
PRPSAP2 | |
Full length recombinant protein of human PRPSAP2 (NP_002758) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1E3 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
MGC117304, MGC126719, MGC126721, PAP4141 kDa phosphoribosypyrophosphate synthetase-associated protein, phosphoribosyl pyrophosphate synthase-associated protein 2, phosphoribosyl pyrophosphate synthetase-associated protein 2, PRPP synthase-associated protein 2 | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
5636.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction