Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRPSAP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531237
Description
Recommended positive controls: K562. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (94%), Chicken (95%), Chimpanzee (98%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The enzyme phosphoribosylpyrophosphate synthetase (PRS) catalyzes the formation of phosphoribosylpyrophosphate which is a substrate for synthesis of purine and pyrimidine nucleotides, histidine, tryptophan and NAD. PRS exists as a complex with two catalytic subunits and two associated subunits. This gene encodes a non-catalytic associated subunit of PRS.
Specifications
PRPSAP2 | |
Polyclonal | |
Unconjugated | |
Prpsap2 | |
41 kDa phosphoribosypyrophosphate synthetase-associated protein; A230054F23Rik; I79_001737; Pap41; phosphoribosyl pyrophosphate synthase-associated protein 2; phosphoribosyl pyrophosphate synthetase associated protein 2; phosphoribosyl pyrophosphate synthetase-associated protein 2; Phosphoribosyl pyrophosphate synthetase-associated protein 2-like protein; PRPP synthase-associated protein 2; PRPP synthetase-associated protein 2; Prpsap2; prpsap2 protein; wu:fi14g04; zgc:55477; zgc:85610 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117272, 5636 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.83 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O08618, O60256 | |
Prpsap2 | |
Recombinant fragment corresponding to a region within amino acids 103 and 359 of Human PRPSAP2. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction