Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRPSAP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554533
Description
Immunogen sequence: VTPPELETKM NITKGGLVLF SANSNSSCME LSKKIAERLG VEMGKVQVYQ EPNRETRVQI QES Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
The enzyme phosphoribosylpyrophosphate synthetase (PRS) catalyzes the formation of phosphoribosylpyrophosphate which is a substrate for synthesis of purine and pyrimidine nucleotides, histidine, tryptophan and NAD. PRS exists as a complex with two catalytic subunits and two associated subunits. This gene encodes a non-catalytic associated subunit of PRS.
Specifications
PRPSAP2 | |
Polyclonal | |
Unconjugated | |
Prpsap2 | |
41 kDa phosphoribosypyrophosphate synthetase-associated protein; A230054F23Rik; I79_001737; Pap41; phosphoribosyl pyrophosphate synthase-associated protein 2; phosphoribosyl pyrophosphate synthetase associated protein 2; phosphoribosyl pyrophosphate synthetase-associated protein 2; Phosphoribosyl pyrophosphate synthetase-associated protein 2-like protein; PRPP synthase-associated protein 2; PRPP synthetase-associated protein 2; Prpsap2; prpsap2 protein; wu:fi14g04; zgc:55477; zgc:85610 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5636 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60256 | |
Prpsap2 | |
Recombinant protein corresponding to Human PRPSAP2. Recombinant protein control fragment (Product #RP-93271). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction