Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PrPSc Antibody (3F4), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281078R
Description
PrPSc Monoclonal specifically detects PrPSc in Human, Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Prion protein | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
PRNP | |
Hamster 263K PrPs. | |
0.1 mL | |
Neuroscience, Stem Cell Markers | |
5621 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
3F4 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CD230, CJD, fatal familial insomnia), GSS, prion protein, prion protein (p27-30), prion protein PrP, prion-related protein, PRIPMGC26679 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Hamster | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction