Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRR11 Antibody (OTI2A10), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273677AF594
Description
PRR11 Monoclonal specifically detects PRR11 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PRR11 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PRR11 | |
Recombinant protein fragment corresponding to amino acids 125-360 of human PRR11 (NP_060774) produced in E.coli. | |
Primary | |
Human, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2A10 | |
Western Blot, Immunohistochemistry | |
FLJ11029, proline rich 11, proline-rich protein 11, transcription repressor of MHCII | |
Mouse | |
0.1 mL | |
55771 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction