Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRR11 Monoclonal Antibody (OTI1A4)
GREENER_CHOICE

Catalog No. PIMA526447
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526447 Supplier Invitrogen™ Supplier No. MA526447
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRR11 Monoclonal Antibody for Western Blot

Plays a critical role in cell cycle progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR11
Applications Western Blot
Classification Monoclonal
Clone OTI1A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRR11
Gene Accession No. Q96HE9
Gene Alias B930067F20Rik; proline rich 11; proline-rich protein 11; PRR11; transcription repressor of MHCII
Gene Symbols PRR11
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 125-360 of PRR11 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55771
Target Species Human, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.