Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRR11 Mouse anti-Human, Clone: OTI1C1, liquid, TrueMAB™
SDP

Catalog No. 501678396
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-8396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-8396 Supplier OriGene Supplier No. TA800542
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Plays a critical role in cell cycle progression.

Specifications

Antigen PRR11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PRR11
Gene Accession No. Q96HE9
Gene Alias proline rich 11
Gene Symbols PRR11
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 125-360 of human PRR11 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55771
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.