Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRR14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565696
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565696 Supplier Invitrogen™ Supplier No. PA565696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IHRTSSTLRRR SRTTPGPEEG PSQKVDRAPQ PTLVVMLEDI ASPRPPAEGF IDETPNFIIP AQRAEPMRIV RQPTPPPGDL EPPFQPSALP ADPLESPPTA PDP Highest antigen sequence identity to the following orthologs - mouse 49%, rat 49%.

The protein encoded by this gene tethers heterochromatin to the nuclear laminar scaffold by binding heterochromatin protein 1 (HP1) and the nuclear lamina. The tether is broken during mitosis and reforms quickly after mitosis, with the encoded protein first binding HP1 and then attaching to the nuclear lamina. This protein also has been shown to promote MyoD activity and skeletal myogenesis. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR14
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRR14
Gene Accession No. Q9BWN1
Gene Alias BC006909; proline rich 14; proline-rich protein 14; PRR14
Gene Symbols PRR14
Host Species Rabbit
Immunogen Recombinant Human PRR14. Recombinant protein control fragment (Product #RP-98686).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78994
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.