Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRR14L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544389
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544389 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544389 Supplier Invitrogen™ Supplier No. PA544389
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DELDGVKAAC PCPQSSPPEQ KEAEPEKRPK KVSQIRIRKT IPRPDPNLTP Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 80%.

The specific function of C22orf30 is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR14L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PRR14L
Gene Accession No. Q5THK1
Gene Alias 6030436E02Rik; C22orf30; C330019G07Rik; Prl14l; proline rich 14 like; proline rich 14-like; proline rich 14-like protein; Protein PRR14L; PRR14L; RGD1305469
Gene Symbols PRR14L
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C22orf30.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253143
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.