Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRR15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570146
Description
This target displays homology in the following species: Cow: 85%; Horse: 92%; Human: 100%; Pig: 85%; Rat: 77%; Sheep: 85%.
PRR15 is a protein coding gene.
Specifications
PRR15 | |
Polyclonal | |
Unconjugated | |
PRR15 | |
P-AF; proline rich 15; proline-rich protein 15; PRR15; RGD1311589 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
222171 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IV56 | |
PRR15 | |
synthetic peptide directed towards the middle region of human PRR15. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction