Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRR15L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554635
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554635 Supplier Invitrogen™ Supplier No. PA554635
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EIGWWKLTFL RKKKSTPKVL YEIPDTYAQT EGDAEPPRPD AGGPNSDFNT RLEKIVDKST KGKHVKVSNS GRFKEKKKVR ATLAENPNLF D Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 88%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR15L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRR15L
Gene Accession No. Q9BU68
Gene Alias ATAD4; ATPase family, AAA domain containing 4; proline rich 15 like; proline rich 15-like; proline-rich protein 15-like protein; Protein ATAD4; PRR15L; testicular tissue protein Li 157
Gene Symbols PRR15L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRR15L. Recombinant protein control fragment (Product #RP-93383).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79170
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.