Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRR16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561919
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561919 Supplier Invitrogen™ Supplier No. PA561919
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KCEDPKRVVP TANPVKTNGT LLRNGGLPGG PNKIPNGDIC CIPNSNLDKA PVQLLMHRPE KDRCPQAGPR ERVRFNEKVQ YHGYCPDCD Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 85%.

Regulator of cell size that promotes cell size increase independently of mTOR and Hippo signaling pathways. Acts by stimulating the translation of specific mRNAs, including those encoding proteins affecting mitochondrial functions. Increases mitochondrial mass and respiration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR16
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRR16
Gene Accession No. Q569H4
Gene Alias 5430406M13Rik; AI607429; DSC54; LARGEN; mesenchymal stem cell protein DSC54; proline rich 16; proline-rich protein 16; Protein Largen; prr16; RGD1564528
Gene Symbols PRR16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRR16. Recombinant protein control fragment (Product #RP-100268).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51334
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.