Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRR19 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PRR19 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PRR19 Polyclonal specifically detects PRR19 in Human samples. It is validated for Western Blot.Specifications
PRR19 | |
Polyclonal | |
Rabbit | |
Human | |
284338 | |
Synthetic peptides corresponding to MGC70924 The peptide sequence was selected from the N terminal of MGC70924. Peptide sequence MDTQGPVSQPFQQPEKPGRVRRRKTRRERNKALVGSRRPLAHHDPPVAIR. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
MGC70924, proline rich 19, proline-rich protein 19 | |
PRR19 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title