Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRR19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567099
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567099 Supplier Invitrogen™ Supplier No. PA567099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DARDAIVHTLQ ACHGCVPDLA LVLRGCQPPL PGAKPGVSER KMTPFWINSP DQVPEQER Highest antigen sequence identity to the following orthologs - mouse 57%, rat 62%.

PRR19 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRR19
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRR19
Gene Accession No. A6NJB7
Gene Alias proline rich 19; proline-rich protein 19; PRR19
Gene Symbols PRR19
Host Species Rabbit
Immunogen Recombinant Human PRR19. Recombinant protein control fragment (Product #RP-107745).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284338
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.